Order Entry
Slovenia
ContactUsLinkComponent
 
Anti-STAT1 Rabbit Polyclonal Antibody
  NOVUNB100-56314
undefined
Anti-STAT1 Rabbit Polyclonal Antibody
  NOVUNB100-56314

 

  • Antibody type:
    Primary
  • Antigen name:
    Signal Transducer And Activator of Transcription 1
  • Antigen symbol:
    STAT1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Western blot:
    Yes
  • Gene ID:
    6772
  • Antigen synonyms:
    STAT91|ISGF-3|IMD31B|CANDF7|IMD31C|IMD31A
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 μg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: STAT1 (signal transducer and activator of transcription 1, 91kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: