Specifications
- Antibody type:Primary
- Antigen name:Gelsolin
- Antigen symbol:GSN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:actin-depolymerizing factor|DKFZp313L0718|AGEL|RP11-477J21.1|brevin|ADF
- Amino acid number:763 to 775
- UniProtKB:P06396
- Sequence:WDDDYWSVDPLDR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Gelsolin (763-775aa WDDDYWSVDPLDR), identical to the related rat sequence, and different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Gelsolin(GSN) detection. Tested with WB, IHC-P, ICC in Human;Mouse;Rat.
Gelsolin also known as GNS is an actin-binding protein that is a key regulator of actin filament assembly and disassembly. Gelsolin is one of the most potent members of the actin-severing gelsolin/villin superfamily. The gene was assigned to human chromosome 9q33.2. Gelsolin is also known as brevin, or actin-depolymerizing factor; it is the principal intracellular and extracellular actin-severing protein. Gelsolin and Gc protein together constitute the extracellular actin-scavenger system which prevents the toxic effects of actin release into the extracellular space under circumstances of cell necrosis. Gelsolin may have therapeutic potential as a mucolytic agent in CF patients. The antiapoptotic activity of gelsolin seems to prevent a step leading to cytochrome c release from the mitochondria into the cytosol.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
WB: The detection limit for GSN is approximately 1.5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Gelsolin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat