Anti-SCAPER Rabbit Polyclonal Antibody
Catalog # AVIVARP39448_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:S-phase cyclin A-associated protein in the ER
- Antigen symbol:SCAPER
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:49855
- Antigen synonyms:ZNF291|MSTP063|Zfp291
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SCAPER (S-phase cyclin A-associated protein in the ER)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: