Anti-EEF2 Rabbit Polyclonal Antibody
BOSSBS-3486R
: Bioss
- Antibody type:Primary
- Antigen name:Elongation factor 2
- Antigen symbol:EEF2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:none
- Antigen synonyms:EF-2|polypeptidyl-tRNA translocase|EEF-2|EF2|elongation factor 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: EEF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse