Orders LinkContactUsLinkComponent
Boxes, with Hinged Lid
Boxes, with Hinged Lid
Supplier:   PLANO W. PLANNET
Boxes, with Hinged Lid
Supplier:  PLANO W. PLANNET
Certificates
Boxes, with Hinged Lid
Boxes, with Hinged Lid
2 Options Available Below
Product Details & Documents
These clear boxes are made of polystyrene.
  • With hinged lid
  • Good chemical resistance
  • Light and stackable
Multipurpose boxes for a wide range of storage and transport applications.
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 25×25×8 mm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 113,00 €
    113,00 €
    1 * 100 items
       113,00 €
  • 50×50×18 mm
    Log in for availability
    Log in for availability
    1 * 50 items
    1 * 50 items - 70,20 €
    70,20 €
    1 * 50 items
       70,20 €