ContactUsLinkComponent
Analytical HPLC columns, XBridge™ Phenyl
Analytical HPLC columns, XBridge™ Phenyl
Supplier:  WATERS
Certificates
Analytical HPLC columns, XBridge™ Phenyl
Supplier:  WATERS
Specifications
  • Catalog No:
  • WATE186003332
  • WATE186003353
  • WATE186003334
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • pH range:
  • 1 - 12
  • 1 - 12
  • 1 - 12
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • USP code:
  • L11
  • L11
  • L11