ContactUsLinkComponent
Reusable half masks, DUETTA
Reusable half masks, DUETTA
Catalog # 111-3051
Supplier:  BARTELS RIEGER
undefined
Reusable half masks, DUETTA
Catalog # 111-3051
Supplier:  BARTELS RIEGER
Specifications
  • Type:
    DUETTA with Bayonet lock
  • Size:
    U*
  • Pk:
    1
Specifications
About this item
Reusable half mask with double bayonet filter connection, plastic mask body with soft, sealing surface and elastic adjustment straps with head gear. Protects against dusts, gases and vapours, or their combinations.
  • Excellent field of vision, unrestricted freedom of movement and outstanding comfort
  • Superior balance through double filter technology
  • Optimal sealing provided by newly designed sealing rim
  • Easily dismantled for cleaning and maintenance
  • One-piece headband for optimal fit
  • Non-allergenic
One mask size fits all face shapes
Certifications: EN 140; 89/686/EWG