Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Cationic Standards
Multi-Element Standards
Internal standard for ICP-OES/MP-AES
Internal standard for ICP-OES/MP-AES
Internal standard for ICP-OES/MP-AES
Catalog # VARI6610030400
Supplier:
VARIAN
CAS Number:
Expand image
Internal standard for ICP-OES/MP-AES
Catalog # VARI6610030400
Supplier:
VARIAN
CAS Number:
Specifications
Pk:
1
Description:
ICP internal standard, 125 ml, contains 100 mg/L
6
Li, Sc, Y, In, Tb and Bi in 5% HNO
3
Specifications
About this item
Product Details
Product Details