Deoxyribonuclease I
Catalog # ROCKMB-101-0500
Supplier: Rockland Immunochemicals
CAS Number:
Specifications
- Pk:0,5 KU
- Enzyme name:Deoxyribonuclease I
- Grade:Molecular biology grade
Specifications
About this item
Deoxyribonuclease I (usually called DNase I), is an endonuclease coded by the human gene DNASE1. DNase I is a nuclease that cleaves DNA preferentially at phosphodiester linkages adjacent to a pyrimidine nucleotide, yielding 5'-phosphate-terminated polynucleotides with a free hydroxyl group on position 3', on average producing tetranucleotides. It acts on single-stranded DNA, double-stranded DNA, and chromatin.