Saltwater aquaculture test kit, FF-3
Catalog # HACH243003
Supplier: Hach
CAS Number:
Specifications
- Pk:1 KIT
- Description:FF-3, Saltwater aquaculture test kit
Specifications
About this item
Test kit used to measures 10 critical parameters with digital titrator, drop count, and colour discs. It is suitable for use in saltwater aquaculture applications.
Delivery: Includes twenty four reagents, one flask, clippers, one graduated cylinder, one colour comparator box, three colour discs, one vial, one delivery tube, one syringe, two plastic tubes, one BOD bottle, one thermometer, one digital titrator, a manual, and a carrying case.