Orders LinkContactUsLinkComponent
Decolorizer, pPb-1
Decolorizer, pPb-1
pPb-6, decolorizer, 10 ml, LeadTrak™, 10 mL
Catalog # HACH2374755
Supplier:   Hach
Decolorizer, pPb-1
Catalog # HACH2374755
Supplier:  Hach
CAS Number:  
Decolorizer, pPb-1
Decolorizer, pPb-1
pPb-6, decolorizer, 10 ml, LeadTrak™, 10 mL
CAS Number:  
Price
42,60 €
Log in for availability
1 * 10 mL
Product Details & Documents
They are used for determining lead by the LeadTrak™ Fast Column Extraction method.
Specifications
Specifications
  • Chemical method:
    LeadTrak™
  • Pk:
    10 mL
  • Description:
    pPb-6, decolorizer, 10 ml
Specifications