Sulphate test kit, SF-1
Catalog # HACH225100
Supplier: Hach
CAS Number:
Specifications
- Pk:1 KIT
- Description:Sulphate test kit, Model SF-1
- Range:50 - 200 mg/l
Specifications
About this item
High concentrations of magnesium or sodium sulphate in water act as cathartics. This test kit provides economical determination of sulphate levels using the Extinction/Turbidimetric methods.
Delivery: Includes graduated cylinder, mixing bottle, reagent, dipstick comparator, instructions and carrying case.