Test kits, tannin and lignin
Catalog # HACH193701
Supplier: Hach
CAS Number:
Specifications
- Pk:1 KIT
- Description:Tannin/lignin test kit, model TA-3
- Range:0 - 15 mg/l
0 - 150 mg/l
Specifications
About this item
Designed to analyse tannin and lignin together and report result as mg/l tannic acid.
- Simple, economical colour disc method
- Easy-to-carry test kit with quick results
Delivery: Kit includes colour disc, comparator box and tubes, reagents, deionized water, mixing bottle, funnel, filter papers, instructions and carrying case.