Orders LinkContactUsLinkComponent
pH test kits, model 17
pH test kits, model 17
Supplier:   Hach
pH test kits, model 17
Supplier:  Hach
Certificates
pH test kits, model 17
4 Options Available Below
Product Details & Documents
It is a simple, low cost colour disc method which is one of the most accurate visual test kit methods for pH.
Delivery information: Kit includes indicator reagent, colour disc, comparator box, two viewing tubes, instructions and carrying case.
Product Family Options
Product Information
  • Range
    Chemical method
    Pk
    Availability
    Price
  • HACH147006
    pH, mid range, model 17F
    pH, mid range, model 17F
    pH 5,6 - 8,4
    Bromothymol blue
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 187,00 €
    187,00 €
    1 * 1 KIT
       187,00 €
  • HACH147008
    pH, mid range, model 17H
    pH, mid range, model 17H
    pH 6,6 - 8,4
    Phenol red
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 187,00 €
    187,00 €
    1 * 1 KIT
       187,00 €
  • HACH147009
    pH, mid range, model 17J
    pH, mid range, model 17J
    pH 7,4 - 9,6
    Thymol blue
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 187,00 €
    187,00 €
    1 * 1 KIT
       187,00 €
  • HACH147011
    pH, wide range, model 17N
    pH, wide range, model 17N
    pH 4,0 - 10,0
    1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT - 206,00 €
    206,00 €
    1 * 1 KIT
       206,00 €
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Colour viewing tube, glass
    Colour viewing tube, glass
    HACH173006
    6
    Log in for availability
    1 * 6 items - 48,80 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Color disc pH, phenol red, 6,6 - 8,4
    Color disc pH, phenol red, 6,6 - 8,4
    HACH9261100
    1
    Log in for availability
    1 * 1 items - 112,00 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Stoppers for glass viewing tube
    Stoppers for glass viewing tube
    HACH173106
    6
    Log in for availability
    1 * 6 items - 19,90 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Colour comparator box
    Colour comparator box
    HACH173200
    1
    Log in for availability
    1 * 1 items - 23,70 €
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Colour viewing tube, plastic with cap
    Colour viewing tube, plastic with cap
    HACH4660004
    4
    Log in for availability
    1 * 4 items - 33,10 €