Visual comparator test kit, ethylene glycol
Catalog # HACH2186400
Supplier: Hach
CAS Number:
Specifications
- Pk:1 KIT
- Description:Ethylene glycol, EG-1 kit
- Range:0 - 150 mg/l
Specifications
About this item
Ethylene glycol (anti-freeze) contamination of automotive lubricants is an indication of a cracked engine block. Glycol presence also can indicate leaks in cooling systems. This test kit model EG-1 presents a purple colour if ethylene glycol and other 1,2 glycols are present in oil or water.
Delivery: Kit includes sample pipettes, vials and reagents for 25 tests, standard solution, instructions and carrying case.