uvex silv-Air Disposable Particle Filtering Respirators, FFP1/FFP2/FFP3
: UVEX
New Product
These respiratory masks feature a seamless headband for a comfortable, secure fit and soft material edges for greater comfort and wearer acceptance. Masks fulfil the requirements of the dolomite dust test. The design ensures compatible fit with uvex eyewear.
- Innovative ori-nasal design of the nose bridge and the soft seal contours to any facial shape
- Exhalation valve for easy airflow exchange which reduces the build-up of heat and moisture inside the mask
- Adjustable nose clip ensures an excellent individual fit
- Upper face seal offers both secure positioning and comfort
- Activated carbon filter, suitable for welding applications
- Soft foam sealing lip reduces the risk of leakage and promotes a proper fit (FFP3 models)
Colour: White
Frequently Bought Together