Orders LinkContactUsLinkComponent
Label Overwraps, Write Protect®
Label Overwraps, Write Protect®
Supplier:   DIVERSIFIED BIOTECH
Label Overwraps, Write Protect®
Certificates
Label Overwraps, Write Protect®
Label Overwraps, Write Protect®
2 Options Available Below
Product Details & Documents
A clear adhesive overwrap to protect writings from organic solvents and caustic agents on labels, surface of microtubes and other laboratory containers.
  • Fits for 0,5, 1,5 and 2,0 ml tubes
  • Adhere to all plastics, glass and metals
  • Temperature range: −196 to +150 °C
Product Family Options
Product Information
  • Colour
    Dimensions
    Availability
    Price
  • DIVEWRIT-1000
    Write protect for labels
    Write protect for labels
    Transparent
    32×13 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 80,40 €
    80,40 €
    1 * 1 Roll
       80,40 €
  • DIVEWRIT-2000
    Write protect for labels
    Write protect for labels
    Transparent
    41×16 mm
    Log in for availability
    Log in for availability
    1 * 1 Roll
    1 * 1 Roll - 83,40 €
    83,40 €
    1 * 1 Roll
       83,40 €