Goggles, uvex flex seal®
DALLS3405X
- Type:Anti-fog, with fabric headband
- Colour:Dark blue
- Seq:02
- Lenses:Clear PC
- Pk:1
Lightweight, low-profile goggles with a silicone body and an optically correct clear polycarbonate lens provide a full field of vision protect the face against impacts and chemical splashes.
- Cured silicone body stays soft and supple in extreme temperatures
- Headband available in fabric or neoprene for chemical applications
- Uvextreme® antifog lens coating and indirect venting system minimises fogging
- Fit over most prescription eyewear
- Economical lens replacement
: ANSI Z87+ and CSA Z94.3 standards