ContactUsLinkComponent
Silica gel, BAKER ANALYZED® 60 - 200 mesh for column chromatography, J.T.Baker®
Silica gel, BAKER ANALYZED® 60 - 200 mesh for column chromatography, J.T.Baker®
Catalog # 3414-01
Supplier:  Avantor
CAS Number:  
undefined
Silica gel, BAKER ANALYZED® 60 - 200 mesh for column chromatography, J.T.Baker®
Catalog # 3414-01
Supplier:  Avantor
CAS Number:  
Specifications
  • Pack type:
    Glass bottle, amber, wide mouth
  • Pk:
    500 g
  • Seq:
    3645
Specifications
Specification Test Results
  • For Laboratory,Research,or Manufacturing Use:
    Passes Test
  • Toluene Absorption:
    Passes Test
  • Loss on Ignition:
    ≤ 10.0 %
  • Alcohol-Ether Soluble Substances:
    ≤ 0.10 %
  • Bulk Density (g/cc), For Information Only:
    = 15 %
  • Mesh - On U.S. No. 60 Sieve:
    ≤ 15 %
  • Mesh - Thru U.S. No. 200 Sieve:
    ≤ 15 %
Specification Test Results
About this item
 :
 :60.08 g/mol
Boiling Pt:2230 °C (1013 hPa)
Melting Pt:1710 °C
Storage Temperature:Ambient
REACH:01-2119379499-16
CAS Number:63231-67-4
 :112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232