ContactUsLinkComponent
VWR®, Rack for Centrifuge Tubes, Dual
VWR®, Rack for Centrifuge Tubes, Dual
  216-0832
undefined
VWR®, Rack for Centrifuge Tubes, Dual
  216-0832
  • For tubes:
    15 and 50 ml
  • No. of Holes:
    2
  • Colour:
    White
  • Seq:
    01
  • Pk:
    5
Polystyrene.
Securely holds 15 and 50 ml centrifuge tubes.