Adapters, receiver, vacuum, Quickfit®
Catalog # BIBBRA11/SC
Supplier: SciLabware
Specifications
- Cone size:14/23 NS
- Socket size:19/26 NS
- Environmentally Preferable:
- Pk:5
Specifications
About this item
PYREX®, borosilicate glass, bent.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Allows up to four flasks to be connected to a condenser outlet. Useful for collecting distillate fractions. Each flask can be progressively filled by rotating the adapter.