Dilatometers, small quartz
Catalog # PERC03190461
Supplier: PerkinElmer
Specifications
- Description:Dilatometer kit with small quartz
- Seq:01
- Pk:1
Specifications
About this item
The kit permits the determination of volumetric changes of materials upon melting or upon cooling from a melt. A typical application would be the determination of shrinkage from a mold.
- Kit includes quartz dilatometer cell, aluminum oxide powder (28 g), quartz sample tube and instruction sheet
- 0,1 and 1,5 ml capacity can be used with the 15 and 28 mm diameter furnace repectively