Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Closures
Chromatography Vial Closures
Caps for diagnostic vials, WHEATON®
Caps for diagnostic vials, WHEATON®
Caps for diagnostic vials, WHEATON®
Catalog # LSUK300180G
Supplier:
DWK Life Sciences
Expand image
Caps for diagnostic vials, WHEATON®
Catalog # LSUK300180G
Supplier:
DWK Life Sciences
Specifications
Cap Size:
15,5 mm
For:
diagnostic vials
Description:
Closure, PP, green, without liner
Pk:
1.000
Specifications
About this item
Product Details
Product Details