Detergent, Trump Supra plus, Ecolab®
Catalog # KAMM9013840
Supplier: KAMMERER
Specifications
- Description:Trump Supra plus, concentrated alkaline detergent
- Environmentally Preferable:
- Packed:1 canister of 12 kg
- Pk:12 kg
Specifications
About this item
Trump Supra plus is a highly concentrated alkaline detergent for use in dishwashing/glassware washing machines.
- Active chlorine for hygienically clean finish
- Powerful cleaning action even at high water hardness
Compatible with materials that are alkaline and bleach resistant - not suitable for aluminium