ContactUsLinkComponent
Dosing pumps for canister
Dosing pumps for canister
Catalog # BODE980029
Supplier:  BODE CHEMIE
Dosing pumps for canister
Catalog # BODE980029
Supplier:  BODE CHEMIE
Specifications
  • Description:
    Dosing pump for 2 l bottle and 5 l canister, with 10 ml stroke
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
Dosing pump with plastic mounts used for various size of bottles and 5 L cans.