Precision tweezers, straight
Supplier: Roth Carl
Certificates
About this item
Magnetic stainless steel (Inox 02 or Inox 08).
- Extremely fine tips
- Ideal for high precision laboratory work, e.g. microscopy, biology and microbiology
- Temperature-resistant up to 400 °C, according to DIN 50914
- Inox 08 tweezers: Excellent resistance to corrosion, good resistance to salt, autoclavable
Frequently Bought Together