Orders LinkContactUsLinkComponent
Forceps
Supplier:   ENTEGRIS
Forceps
Supplier:  ENTEGRIS
Certificates
5 Options Available Below
Product Details & Documents
ETFE, white. Durable forceps for substrate handling.
  • Different tip styles available
  • Packaged in protective cases
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Straight, 0,3 mm thick tip
    124 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
    124 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • Straight, rounded tip (0,8 mm radius)
    127 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • Curved, tapered thin tip
    127 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items
  • Straight, rounded tip (1,5 mm radius, 3,0 mm thickness)
    178 mm
    Log in for availability
    Log in for availability
    1 * 4 items
    1 * 4 items