ContactUsLinkComponent
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Catalog # 84960.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Catalog # 84960.180
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Chlorate standard solution
  • Contents:
    NaClO₃ in H₂O
Specifications
About this item
Certifications: Traceable to NIST. Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities. Supplied with Certificate of Analysis.