Orders LinkContactUsLinkComponent
Labelling pen, quick drying
Labelling pen, quick drying
Labelling pen with fine, Black
Catalog # BRDY335091
Supplier:   Brady
Labelling pen, quick drying
Catalog # BRDY335091
Supplier:  Brady
Labelling pen, quick drying
Labelling pen, quick drying
Labelling pen with fine, Black
Price
26,40 €
Log in for availability
1 * 5 items
Product Details & Documents
These labelling pens use a quick drying ink.
The permanent ink is specially formulated to resist oil, solvents, abrasion and fading.
Specifications
  • Colour:
    Black
  • Description:
    Labelling pen with fine
  • Seq:
    02