ContactUsLinkComponent
Dispenser for protective laboratory labelling tapes, SP Bel-Art
Dispenser for protective laboratory labelling tapes, SP Bel-Art
Catalog # 817-1130
undefined
Dispenser for protective laboratory labelling tapes, SP Bel-Art
Catalog # 817-1130
Specifications
  • Colour:
    Transparent
  • Description:
    Dispenser for labelling tape
  • Pk:
    1
Specifications
About this item
The tape dispenser is moulded from strong ABS plastic with a serrated steel cutting blade.
  • Tape roll held in place by aluminum roller and spring-loaded cover