ContactUsLinkComponent
Surface disinfectants, perform® sterile alcohol IPA
  141-1462
undefined
Surface disinfectants, perform® sterile alcohol IPA
  141-1462
  • Description:
    perform® sterile alcohol IPA, surface disinfectant,
  • Packed:
    500 ml × 10 spray bottles per case
  • Pk:
    10 Bottle
Sterile, ready to use alcohol-based solution for surface disinfection in cleanrooms.
  • Fast and effective
  • Gamma irradiated and microbial filtered (0,2 μm)
  • Colour, perfume and residue free
  • Bactericidal, virucidal (BVDV/Vaccinia), fungicidal
 : Use biocides safely. Always read the label and product information before use.