ContactUsLinkComponent
Accessories for Kjeldahl digestion units, automatic, DKL series
Accessories for Kjeldahl digestion units, automatic, DKL series
Catalog # 705-0863
Supplier:  VELP SCIENTIFIC
undefined
Accessories for Kjeldahl digestion units, automatic, DKL series
Catalog # 705-0863
Supplier:  VELP SCIENTIFIC
Specifications
  • Description:
    Test tube, 400 ml, Ø 50×300 mm, for DKL 20 and DKL 12, 1 piece/box
  • Seq:
    25
  • Pk:
    1