Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Controllers
Temperature Controllers
Accessories for digital temperature controller, MC8108
Accessories for digital temperature controller, MC8108
Accessories for digital temperature controller, MC8108
Catalog # ELET99965-02
Supplier:
Cole-Parmer
Expand image
Accessories for digital temperature controller, MC8108
Catalog # ELET99965-02
Supplier:
Cole-Parmer
Specifications
Description:
Temperature probe, max. 250 °C, stainless steel with PTFE sleeving
Seq:
01
Pk:
1
Specifications
About this item
Product Details
Product Details