ContactUsLinkComponent
Sleeve protector, PP/PE, Sekuroka®
Sleeve protector, PP/PE, Sekuroka®
Catalog # ROTH8202.2
Supplier:  Roth Carl
Sleeve protector, PP/PE, Sekuroka®
Catalog # ROTH8202.2
Supplier:  Roth Carl
Specifications
  • Garment application:
    Laboratory
  • Colour:
    White
  • Length:
    580 mm
  • Pk:
    10 PAIR
Specifications
About this item
These protective sleeves with elastic bands are made of PP with PE coating.
  • Anti-static, lint-free
  • Tearproof and water repellent