Microtubes with cap, 1,5 ml
1,5 ml, Natural, with attached lid, graduated, conical bottom, non sterile
211-2081
: SEMADENI
PP, translucent.
- Pierceable lid
- Autoclavable
- Withstand centrifugation at 21000×g
- Excellent chemical resistance
With attached push-on lid.
- Type:with attached lid
- Version:graduated, conical bottom, non sterile
- Capacity:1,5 ml
- Colour:Natural
- Seq:01