Orders LinkContactUsLinkComponent
Trays, multipurpose, PP
Trays, multipurpose, PP
Supplier:   SEMADENI
Trays, multipurpose, PP
Supplier:  SEMADENI
Certificates
Trays, multipurpose, PP
Trays, multipurpose, PP
4 Options Available Below
Product Details & Documents
Made of PP, white.
  • Stable, durable, unbreakable
  • Temperature resistant from −20 to +130 °C
  • Smooth surfaces for easy cleaning
Multipurpose trays. Suitable for use as a tray for aggressive substances, sterilisation and photographic development.
Documents
Product Family Options
Product Information
  • Colour
    Material
    W×D×H
    Availability
    Price
  • White
    PP
    250×310×65 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 22,10 €
    22,10 €
    1 * 1 items
       22,10 €
  • White
    PP
    310×370×75 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 31,60 €
    31,60 €
    1 * 1 items
       31,60 €
  • White
    PP
    420×520×120 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 56,10 €
    56,10 €
    1 * 1 items
       56,10 €
  • White
    PP
    535×635×140 mm
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 104,00 €
    104,00 €
    1 * 1 items
       104,00 €