Gowning room wipes, pre-saturated, PROSAT®
Catalog # CONT497600-960_U
Supplier: CONTEC
Specifications
- Packed:800 wipes/bag
- L×W:130×200 mm
- Seq:01
- Pk:800
Specifications
About this item
These non woven wipes are made from strong, lint-free cellulose/polyester fabric, saturated with 6% IPA and 94% DI water. They are ideal for glove wipe down during gowning procedures.
- Easy access dispensing tub
- Very low residue
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904