UPLC columns, ACQUITY UPLC™ BEH Shield RP 18
Supplier: WATERS
Certificates
About this item
These columns complement the ACQUITY BEH C18 and C8 phases and provide a necessary and powerful tool for HPLC method development. The embedded carbamate group in the bonded phase ligand provides alternate selectivity, especially for phenolic compounds compared to straight chain alkyl columns.
Specifications
- Catalog No:
- WATE186002853
- WATE186002854
- Carbon load:
- 17%
- 17%
- Endcapped:
- Yes, Yes
- Yes, Yes
- pH range:
- 2 - 11
- 2 - 11
- Surface area:
- 185 m²/g
- 185 m²/g
- USP code:
- L1, L1
- L1, L1