ContactUsLinkComponent
UPLC columns, ACQUITY UPLC™ BEH phenyl
UPLC columns, ACQUITY UPLC™ BEH phenyl
Supplier:  WATERS
Certificates
UPLC columns, ACQUITY UPLC™ BEH phenyl
Supplier:  WATERS
Certificates
Specifications
  • Catalog No:
  • WATE186002884
  • WATE186003911
  • WATE186002885
  • WATE186003375
  • Carbon load:
  • 15%
  • 15%
  • 15%
  • 15%
  • Endcapped:
  • Yes, Yes
  • Yes, Yes
  • Yes, Yes
  • Yes, Yes
  • pH range:
  • 1 - 12
  • 1 - 12
  • 1 - 12
  • 1 - 12
  • Surface area:
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • 185 m²/g
  • USP code:
  • L11, L11
  • L11, L11
  • L11, L11
  • L11, L11