Orders LinkContactUsLinkComponent
Measuring scoops, PP
 :   SEMADENI
Measuring scoops, PP
 :  SEMADENI
Measuring scoops, PP
Measuring scoops, PP
PP, white. Shatterproof measuring scoops with good chemical resistance.
  • With flat, bevelled end and reinforced handle
  • Heat resistant up to 130 °C, sterilisable
  • Volume is visible on the handle of each scoop
The scoops are also food contact approved and can therefore be used for food manufacturing or processing applications.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
  • Capacity
    Length
    Width
    Colour
  • 5 ml
    82 mm
    25 mm
    White
    1 * 1 items
    1 * 1 items - 1,80 €
    1,80 €
    1 * 1 items
       1,80 €
  • 10 ml
    100 mm
    35 mm
    White
    1 * 1 items
    1 * 1 items - 2,30 €
    2,30 €
    1 * 1 items
       2,30 €
  • 25 ml
    135 mm
    45 mm
    White
    1 * 1 items
    1 * 1 items - 2,60 €
    2,60 €
    1 * 1 items
       2,60 €
  • 50 ml
    160 mm
    55 mm
    White
    1 * 1 items
    1 * 1 items - 3,40 €
    3,40 €
    1 * 1 items
       3,40 €
  • 100 ml
    200 mm
    65 mm
    White
    1 * 1 items
    1 * 1 items - 4,90 €
    4,90 €
    1 * 1 items
       4,90 €
  • 250 ml
    260 mm
    90 mm
    White
    1 * 1 items
    1 * 1 items - 11,20 €
    11,20 €
    1 * 1 items
       11,20 €
  • 500 ml
    315 mm
    110 mm
    White
    1 * 1 items
    1 * 1 items - 15,20 €
    15,20 €
    1 * 1 items
       15,20 €
  • 1000 ml
    385 mm
    150 mm
    White
    1 * 1 items
    1 * 1 items - 17,60 €
    17,60 €
    1 * 1 items
       17,60 €
This product has been blocked by your organization. Please contact your purchasing department for more information