Orders LinkContactUsLinkComponent
Multi-parameter meters, handheld, 3320
Supplier:   WTW
Multi-parameter meters, handheld, 3320
Supplier:  WTW
Certificates
Multi-parameter meters, handheld, 3320
5 Options Available Below
Product Details & Documents
Multi 3320: This advanced meter features two inputs for the simultaneous measurement of pH and conductivity, or pH and dissolved oxygen. It is perfect for measurements in ground and surface water, aquacultures and wastewater.
  • Backlit LCD for displaying two measurements at the same time
  • Robust and waterproof housing (IP 67)
  • Memory with data logging functionality between 1 s and 60 min
  • All entries are GLP compliant with time, date and selectable ID number
  • Waterproof USB interface enables fast and easy data transfer
pH/Cond 3320: This portable meter features two inputs for the simultaneous measurement of conductivity and pH or ISE or ORP. It is ideal for almost every application in process chemistry, life science, food and beverage applications as well as pharmaceutical industry (measurement of pH and conductivity according to pharmacopeia standards).
This advanced meter features two inputs for the simultaneous measurement of pH and conductivity, or pH and dissolved oxygen. It is perfect for measurements in ground and surface water, aquacultures and waste water.
 : Single instruments are supplied with manual, CD-ROM, batteries and USB cable. The meters are also available in sets including sensors and accessories.
Specifications for Product Family
Specifications
  • Catalog No:
  • 665-0313
  • 665-0315
  • 665-0319
  • 665-0320
  • 665-0321
  • Model:
  • pH/Cond 3320
  • pH/Cond 3320
  • Multi 3320
  • Multi 3320
  • Multi 3320
  • pH range:
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • –2,000...+19,999
  • mV range:
  • ±2500
  • ±2500
  • ±2500
  • ±2500
  • ±2500
  • ISE range:
  • 0,000...999999
  • 0,000...999999
  • 0,000...999999
  • 0,000...999999
  • 0,000...999999
  • ISE units:
  • mg/l, µmol/l, mg/kg, ppm, %
  • mg/l, µmol/l, mg/kg, ppm, %
  • mg/l, µmol/l, mg/kg, ppm, %
  • mg/l, µmol/l, mg/kg, ppm, %
  • mg/l, µmol/l, mg/kg, ppm, %
  • Conductivity range:
  • 0,000 µS/cm - 1000 mS/cm
  • 0,000 µS/cm - 1000 mS/cm
  • 0,000 µS/cm - 1000 mS/cm
  • 0,000 µS/cm - 1000 mS/cm
  • 0,000 µS/cm - 1000 mS/cm
  • Conductivity reference temperature:
  • 20/25 °C
  • 20/25 °C
  • 20/25 °C
  • 20/25 °C
  • 20/25 °C
  • Salinity range:
  • 0,0 - 70,0 ppt
  • 0,0 - 70,0 ppt
  • 0,0 - 70,0 ppt
  • 0,0 - 70,0 ppt
  • 0,0 - 70,0 ppt
  • TDS range:
  • 0 - 199,9 g/l
  • 0 - 199,9 g/l
  • 0 - 199,9 g/l
  • 0 - 199,9 g/l
  • 0 - 199,9 g/l
  • DO concentration:
  • -
  • -
  • 0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
  • 0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
  • 0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
  • DO saturation:
  • -
  • -
  • 0,00 - 200,0; 0,0 - 600 % (depending on sensor)
  • 0,00 - 200,0; 0,0 - 600 % (depending on sensor)
  • 0,00 - 200,0; 0,0 - 600 % (depending on sensor)
  • Display:
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Graphic LCD, backlit
  • Power supply:
  • 4×1,5 V AA batteries, 4×1,2 V NiMH Akku or USB
  • 4×1,5 V AA batteries, 4×1,2 V NiMH Akku or USB
  • 4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
  • 4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
  • 4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
  • Compliance:
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • IP 67
  • Data logging - log function:
  • Manual/time controlled
  • Manual/time controlled
  • Manual/time controlled
  • Manual/time controlled
  • Manual/time controlled
  • Data logging capacity:
  • 5000 data sets (automatic); 200 data sets (manual)
  • 5000 data sets (automatic); 200 data sets (manual)
  • 5000 data sets (automatic); 200 data sets (manual)
  • 5000 data sets (automatic); 200 data sets (manual)
  • 5000 data sets (automatic); 200 data sets (manual)
  • Connections:
  • Mini-USB
  • Mini-USB
  • Mini-USB
  • Mini-USB
  • Mini-USB
  • W×D×H:
  • 80×55×180 mm
  • 80×55×180 mm
  • 80×55×180 mm
  • 80×55×180 mm
  • 80×55×180 mm
  • Weight:
  • 0,4 kg
  • 0,4 kg
  • 0,4 kg
  • 0,4 kg
  • 0,4 kg
Specifications
Product Family Options
pH/Cond3320
Product Information
  • Availability
    Price
  • 665-0313
    pH/Cond 3320, single instrument, incl. manual, CD-ROM, batteries and cable
    pH/Cond 3320, single instrument, incl. manual, CD-ROM, batteries and cable
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 340,00 €
    1 340,00 €
    1 * 1 items
       1 340,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    665-0313
    Model
    pH/Cond 3320
    pH range
    –2,000...+19,999
    mV range
    ±2500
    ISE range
    0,000...999999
    ISE units
    mg/l, µmol/l, mg/kg, ppm, %
    Conductivity range
    0,000 µS/cm - 1000 mS/cm
    Conductivity reference temperature
    20/25 °C
    Salinity range
    0,0 - 70,0 ppt
    TDS range
    0 - 199,9 g/l
    DO concentration
    -
    DO saturation
    -
    Display
    Graphic LCD, backlit
    Power supply
    4×1,5 V AA batteries, 4×1,2 V NiMH Akku or USB
    Compliance
    IP 67
    Data logging - log function
    Manual/time controlled
    Data logging capacity
    5000 data sets (automatic); 200 data sets (manual)
    Connections
    Mini-USB
    W×D×H
    80×55×180 mm
    Weight
    0,4 kg
  • 665-0315
    pH/Cond 3320, set 2, incl. instrument, carrying case, sensors SenTix® 41 and TetraCon® 325, buffer solutions STP 4 and STP 7, conductivity standard 0,01 mol/L KCl, manual, stand, beaker, CD-ROM, batteries and cable
    pH/Cond 3320, set 2, incl. instrument, carrying case, sensors SenTix® 41 and TetraCon® 325, buffer solutions STP 4 and STP 7, conductivity standard 0,01 mol/L KCl, manual, stand, beaker, CD-ROM, batteries and cable
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 940,00 €
    1 940,00 €
    1 * 1 items
       1 940,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    665-0315
    Model
    pH/Cond 3320
    pH range
    –2,000...+19,999
    mV range
    ±2500
    ISE range
    0,000...999999
    ISE units
    mg/l, µmol/l, mg/kg, ppm, %
    Conductivity range
    0,000 µS/cm - 1000 mS/cm
    Conductivity reference temperature
    20/25 °C
    Salinity range
    0,0 - 70,0 ppt
    TDS range
    0 - 199,9 g/l
    DO concentration
    -
    DO saturation
    -
    Display
    Graphic LCD, backlit
    Power supply
    4×1,5 V AA batteries, 4×1,2 V NiMH Akku or USB
    Compliance
    IP 67
    Data logging - log function
    Manual/time controlled
    Data logging capacity
    5000 data sets (automatic); 200 data sets (manual)
    Connections
    Mini-USB
    W×D×H
    80×55×180 mm
    Weight
    0,4 kg
Multi 3320
Product Information
  • Availability
    Price
  • 665-0319
    Multi 3320, single instrument, incl. manual, CD-ROM, batteries and cable
    Multi 3320, single instrument, incl. manual, CD-ROM, batteries and cable
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 1 720,00 €
    1 720,00 €
    1 * 1 items
       1 720,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    665-0319
    Model
    Multi 3320
    pH range
    –2,000...+19,999
    mV range
    ±2500
    ISE range
    0,000...999999
    ISE units
    mg/l, µmol/l, mg/kg, ppm, %
    Conductivity range
    0,000 µS/cm - 1000 mS/cm
    Conductivity reference temperature
    20/25 °C
    Salinity range
    0,0 - 70,0 ppt
    TDS range
    0 - 199,9 g/l
    DO concentration
    0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
    DO saturation
    0,00 - 200,0; 0,0 - 600 % (depending on sensor)
    Display
    Graphic LCD, backlit
    Power supply
    4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
    Compliance
    IP 67
    Data logging - log function
    Manual/time controlled
    Data logging capacity
    5000 data sets (automatic); 200 data sets (manual)
    Connections
    Mini-USB
    W×D×H
    80×55×180 mm
    Weight
    0,4 kg
  • 665-0320
    Multi 3320, set 1, incl. instrument, field case, sensors SenTix® 41, CellOx® 325, TetraCon® 325, manual, stand, beaker, CD-ROM, batteries and cable
    Multi 3320, set 1, incl. instrument, field case, sensors SenTix® 41, CellOx® 325, TetraCon® 325, manual, stand, beaker, CD-ROM, batteries and cable
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3 230,00 €
    3 230,00 €
    1 * 1 items
       3 230,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    665-0320
    Model
    Multi 3320
    pH range
    –2,000...+19,999
    mV range
    ±2500
    ISE range
    0,000...999999
    ISE units
    mg/l, µmol/l, mg/kg, ppm, %
    Conductivity range
    0,000 µS/cm - 1000 mS/cm
    Conductivity reference temperature
    20/25 °C
    Salinity range
    0,0 - 70,0 ppt
    TDS range
    0 - 199,9 g/l
    DO concentration
    0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
    DO saturation
    0,00 - 200,0; 0,0 - 600 % (depending on sensor)
    Display
    Graphic LCD, backlit
    Power supply
    4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
    Compliance
    IP 67
    Data logging - log function
    Manual/time controlled
    Data logging capacity
    5000 data sets (automatic); 200 data sets (manual)
    Connections
    Mini-USB
    W×D×H
    80×55×180 mm
    Weight
    0,4 kg
  • 665-0321
    Multi 3320, set 2, incl. instrument, field case, sensors SenTix® 41-3, CellOx® 325-3 and TetraCon® 325-3 (all with 3 m cable), manual, stand, beaker, CD-ROM, batteries and cable
    Multi 3320, set 2, incl. instrument, field case, sensors SenTix® 41-3, CellOx® 325-3 and TetraCon® 325-3 (all with 3 m cable), manual, stand, beaker, CD-ROM, batteries and cable
    Log in for availability
    Log in for availability
    1 * 1 items
    1 * 1 items - 3 360,00 €
    3 360,00 €
    1 * 1 items
       3 360,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    665-0321
    Model
    Multi 3320
    pH range
    –2,000...+19,999
    mV range
    ±2500
    ISE range
    0,000...999999
    ISE units
    mg/l, µmol/l, mg/kg, ppm, %
    Conductivity range
    0,000 µS/cm - 1000 mS/cm
    Conductivity reference temperature
    20/25 °C
    Salinity range
    0,0 - 70,0 ppt
    TDS range
    0 - 199,9 g/l
    DO concentration
    0,00 - 20,00 / 0,0 - 90,0 mg/L (depending on sensor)
    DO saturation
    0,00 - 200,0; 0,0 - 600 % (depending on sensor)
    Display
    Graphic LCD, backlit
    Power supply
    4 × 1,5 V AA batteries, 4 × 1,2 V NiMH Akku or USB
    Compliance
    IP 67
    Data logging - log function
    Manual/time controlled
    Data logging capacity
    5000 data sets (automatic); 200 data sets (manual)
    Connections
    Mini-USB
    W×D×H
    80×55×180 mm
    Weight
    0,4 kg