Orders LinkContactUsLinkComponent
Slide container
Slide container
Slide containers, Translucent, HDPE, 2
Catalog # 212-0530
Supplier:   BOETTGER PAUL
Slide container
Catalog # 212-0530
Supplier:  BOETTGER PAUL
Slide container
Slide container
Slide containers, Translucent, HDPE, 2
Price
135,00 €
Log in for availability
1 * 500 items
Product Details & Documents
High-density polyethylene container for microscopy slides.
  • Mailing or storage of microscopy slides
  • For two slides (or four slides stored opposite each other)
  • Shielded ventilation in the cover
Ordering information: Containers are supplied without slides.
Specifications
  • No. of slides:
    2
  • Colour:
    Translucent
  • Description:
    Slide containers
  • Material:
    HDPE