ContactUsLinkComponent
VWR® Q Path® plus Adhesion Slides
VWR® Q Path® plus Adhesion Slides
Catalog # 630-2092
Supplier:  LABONORD
undefined
VWR® Q Path® plus Adhesion Slides
Catalog # 630-2092
Supplier:  LABONORD
Specifications
  • Colour:
    White
  • Description:
    Adhesion slides, Q Path® plus
  • Pk:
    72
Specifications
About this item
These slides can be used for the diagnosis of suspensions of cells and samples of tissue and are suitable for in vitro diagnostics.
Certifications: ISO 8037/1. EN 375: 2001.