Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Environmental Test Kits
Visual comparator disc, phosphate
Visual comparator disc, phosphate
Catalog # HACH9262100
Supplier:
Hach
CAS Number:
Expand image
Visual comparator disc, phosphate
Catalog # HACH9262100
Supplier:
Hach
CAS Number:
Specifications
Pk:
1
Description:
Phosphate, colour disc
Range:
0 - 40 mg/l PO₄
Specifications
About this item
Product Details
Fast acccurate comparison
Product Details