Orders LinkContactUsLinkComponent
Mailing container, with screw cap, square
Mailing container, with screw cap, square
Supplier:   SARSTEDT
Mailing container, with screw cap, square
Supplier:  SARSTEDT
Certificates
Mailing container, with screw cap, square
2 Options Available Below
Product Details & Documents
Containers for safe postage and transportation of clinical samples (blood, urine, swabs and faeces tubes).
  • Tight screw caps ensures the safety
  • Made of neutral HDPE
  • One side of each container can be printed
  • Available with or without absorbent liners
Suitable for 30 ml liquid sample and faeces tubes of 107ר25 mm.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • SARS78.572
    Mailing container, square, 117×35 mm, Ø opening: 28 mm, with cap
    Mailing container, square, 117×35 mm, Ø opening: 28 mm, with cap
    Natural
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - 89,70 €
    89,70 €
    1 * 250 items
       89,70 €
  • SARS78.573
    Mailing container with absorbent liner, square, 117×35 mm, Ø opening: 28 mm, with cap
    Mailing container with absorbent liner, square, 117×35 mm, Ø opening: 28 mm, with cap
    Natural
    Log in for availability
    Log in for availability
    1 * 250 items
    1 * 250 items - 123,00 €
    123,00 €
    1 * 250 items
       123,00 €