Caps for screw neck vials
WATE186000305
: WATERS
- For:Alliance systems
- Septum:blue PTFE/white silicone, pre-slit
- Type:Screw cap
- Pk:100
Dark blue PE screw cap with bonded slit septum for 12×32 mm vials.
Dark blue PE screw cap with bonded slit septum for 12×32 mm vials.