Storage boxes
Supplier: MARIENFELD
Jump to:
About this item
Plastic boxes for microscope slides.
- Dust-tight securely snap-on lid
- Enclosed index card
- Numbered slots inside
Designed for the safe storage of slides during transport.
Slide size: 76×26 mm
2 Options Available Below
Product Details & Documents
Plastic boxes for microscope slides.
- Dust-tight securely snap-on lid
- Enclosed index card
- Numbered slots inside
Designed for the safe storage of slides during transport.
Slide size: 76×26 mm
Product Family Options
Product Information
- ColourMaterialNo. of slidesW×D×HAvailabilityPrice
- MARI5620020Storage boxes
- MARI5620028Storage box, stackable, with dust-tight snap-on lidStorage box, stackable, with dust-tight snap-on lidWhiteABS50— mmLog in for availability1 * 1 items11,42 €