ContactUsLinkComponent
Rack for pipetting pumps
Rack for pipetting pumps
Catalog # 612-2080
Supplier:  DELTALAB
Rack for pipetting pumps
Catalog # 612-2080
Supplier:  DELTALAB
Specifications
  • For:
    4 pipette pumps (W-100/W-110/W-120)
  • Colour:
    Transparent
  • Description:
    Support rack
  • W×D×H:
    109×208×144 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Pipette pump support rack made of acrylic resin and designed to keep in position four pipette pumps.
  • Angled for quick placement and removal of pipettors
  • U-shaped channels and a lower deck provide secure and convenient storage
  • Suitable for pipette pumps W-100, W-110, W-120