ContactUsLinkComponent
Racks for microcentrifuge tubes
Racks for microcentrifuge tubes
Catalog # DELT200420
Supplier:  DELTALAB
Racks for microcentrifuge tubes
Catalog # DELT200420
Supplier:  DELTALAB
Specifications
  • For tubes:
    1,5/2,0 ml
  • No. of Holes:
    20
  • Colour:
    Blue
  • W×D×H:
    210×90×48 mm
  • Environmentally Preferable:
  • Pk:
    6
Specifications
About this item
PP, Two level.
  • Interlocking - can be extended lengthways
  • Two levels for easy manipulation of the tubes
  • Easy to read numbers for better tube viewing and identification.