ContactUsLinkComponent
Safety knives for left and right-handed users, SECUNORM PROFI
Safety knives for left and right-handed users, SECUNORM PROFI
Catalog # 233-2245
Supplier:  MARTOR ARGENTAX
undefined
Safety knives for left and right-handed users, SECUNORM PROFI
Catalog # 233-2245
Supplier:  MARTOR ARGENTAX
Specifications
  • Type:
    For right-handed users
  • Total length:
    142 mm
  • Blade:
    53×19×0,63 mm
  • Pk:
    1
Specifications
About this item
Trapezoid blade, aluminium handle.
  • Versions available for left or right-handed users
  • When cutting, the user must remove their thumb from the slider
  • Easy blade replacement by sliding back the inner part, without the need for tools
These classic safety knives have a spring-loaded blade which automatically disappears into the handle if the blade slips off the material being cut and the slider is not held down. They are especially suitable for slitting cardboard boxes and adhesive tape.
TÜV tested
Certifications: TÜV tested